Order Entry
Poland
ContactUsLinkComponent
2-Methoxy-4-[(E)-2-nitrovinyl]phenol 98%
Irritant
2-Methoxy-4-[(E)-2-nitrovinyl]phenol 98%
Catalog # APOSOR26919-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methoxy-4-[(E)-2-nitrovinyl]phenol 98%
Catalog # APOSOR26919-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :195.172 g/mol
Melting Pt:168…170 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:6178-42-3
Formula:C₉H₉NO₄
MDL Number:MFCD00024827