1-(4-Chlorophenyl)-3-hydroxy-1,2-dihydroquinoxalin-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:74769-78-1
:
Formula:C₁₄H₉ClN₂O₂
MDL Number:MFCD00100500
:272.686 g/mol
Storage Temperature:Ambient
Frequently Bought Together