4,6-Dibromobenzene-1,3-diamine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1,5-Diamino-2,4-dibromobenzene
CAS Number:36210-57-8
:
Formula:C₆H₆Br₂N₂
MDL Number:MFCD22042731
Molecular Weight:265.933 g/mol
Melting Pt:133…135 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together