Order Entry
Poland
ContactUsLinkComponent
3-(4-Acetamidophenyl)-1H-pyrazole
3-(4-Acetamidophenyl)-1H-pyrazole
Catalog # APOSOR183452-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Acetamidophenyl)-1H-pyrazole
Catalog # APOSOR183452-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.225 g/mol
Storage Temperature:Ambient
CAS Number:197093-26-8
Formula:C₁₁H₁₁N₃O
MDL Number:MFCD09028475