Order Entry
Poland
ContactUsLinkComponent
3-Bromo-7-methyl-1H-indole, N-BOC protected 95+%
Corrosive
Irritant
3-Bromo-7-methyl-1H-indole, N-BOC protected 95+%
Catalog # APOSOR1741-500MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
3-Bromo-7-methyl-1H-indole, N-BOC protected 95+%
Catalog # APOSOR1741-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :310.186 g/mol
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:914349-39-6
Formula:C₁₄H₁₆BrNO₂
MDL Number:MFCD05864788