1'-[(1-Hydroxy-1H-benzotriazol-6-yl)sulphonyl]-1,4'-bipiperidine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:227471-63-8
:
Formula:C₁₆H₂₃N₅O₃S
MDL Number:MFCD04114694
:365.451 g/mol
Melting Pt:195…200 °C
Storage Temperature:Ambient
Frequently Bought Together