2-Methylpiperazine, N1-CBZ protected 96%
Catalog # APOSOR16019-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:234.294 g/mol
Storage Temperature:Ambient
CAS Number:444666-46-0
Formula:C₁₃H₁₈N₂O₂
MDL Number:MFCD04115058