Order Entry
Poland
ContactUsLinkComponent
2-(2-Chloro-3,4-dimethoxybenzyl)-1H-isoindole-1,3(2H)-dione
2-(2-Chloro-3,4-dimethoxybenzyl)-1H-isoindole-1,3(2H)-dione
Catalog # APOSOR111608-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Chloro-3,4-dimethoxybenzyl)-1H-isoindole-1,3(2H)-dione
Catalog # APOSOR111608-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :331.75 g/mol
Storage Temperature:Ambient
CAS Number:2227107-24-4
Formula:C₁₇H₁₄ClNO₄
MDL Number:MFCD31621196