Ethyl 4-(4-chlorophenyl)-3-methyl-2,4-dioxobutanoate
Catalog # APOSOR111464-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:268.693 g/mol
Storage Temperature:Ambient
CAS Number:169544-41-6
Formula:C₁₃H₁₃ClO₄
MDL Number:MFCD06411133