
1-(Oxolan-2-ylmethyl)-1H-pyrazol-4-amine
Catalog # APOSOR111069-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.208 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1029413-45-3
Formula:C₈H₁₃N₃O
MDL Number:MFCD11128191