N-(tert-Butyl)-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Catalog # APOSOR110845-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.26 g/mol
Storage Temperature:Ambient
CAS Number:869072-45-7
Formula:C₁₀H₁₅N₅
MDL Number:MFCD03180509