4-Hydrazino-5-thien-2-ylthieno[2,3-d]pyrimidine
Catalog # APOSOR110641-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:248.327 g/mol
Storage Temperature:Ambient
CAS Number:406200-90-6
Formula:C₁₀H₈N₄S₂
MDL Number:MFCD03444393