
4-Cyclohexylpiperazine-1-carboximidamide hydroiodide
Catalog # APOSOR110027-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:338.232 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:849776-37-0
Formula:C₁₁H₂₂N₄·HI
MDL Number:MFCD07368564