Order Entry
Poland
ContactUsLinkComponent
2-(4-Methoxybenzoyl)indane-1,3-dione 98%
Irritant
2-(4-Methoxybenzoyl)indane-1,3-dione 98%
Catalog # APOSOR1032-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(4-Methoxybenzoyl)indane-1,3-dione 98%
Catalog # APOSOR1032-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :280.275 g/mol
Melting Pt:123…124 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:147847-17-4
Formula:C₁₇H₁₂O₄
MDL Number:MFCD00129403