
6-Methoxy-7,8,9,10-tetrahydro-6H-1,2 5-oxadiazolo[3 4-c]carbazole
Catalog # APOSOR0590-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:243.261 g/mol
Melting Pt:118…120 °C
:GHS07
Storage Temperature:Ambient
CAS Number:255865-29-3
Formula:C₁₃H₁₃N₃O₂
MDL Number:MFCD00168557