Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-Urocanic acid-[13C3]
cis-Urocanic acid-[13C3]
cis-Urocanic acid-[13C3]
cis-Urocanic acid-[13C3]
Catalog # APOSBISC1057-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
cis-Urocanic acid-[13C3]
Catalog # APOSBISC1057-1MG
Supplier:
Apollo Scientific
CAS Number:
About this item
Product Details
Product Details