Order Entry
Poland
ContactUsLinkComponent
Disinfectant, Divosan Forte VT6
Disinfectant, Divosan Forte VT6
Catalog # DIVY7510702
Supplier:  Diversey
Disinfectant, Divosan Forte VT6
Catalog # DIVY7510702
Supplier:  Diversey
Specifications
  • Description:
    Divosan Forte VT6, disinfectant
  • Seq:
    02
  • Pk:
    200 L
Specifications
About this item
Divosan Forte is a high-activity oxidising disinfectant based on peracetic acid for use in the food, beverage and dairy industries. Divosan Forte is a stabilised peracetic acid solution (15%) which is nonfoaming and completely free-rinsing.
  • Highly effective disinfectant against all types of microorganisms
  • Terminal disinfectant for use in automated CIP systems
  • Excellent deodorising and stain removal properties
  • Automatic injection using suitable CIP dosing equipment
  • Highly concentrated product for optimum cost-effectiveness
  • Suitable for use in soft or hard water
Versatile and effective CIP disinfectant, can be used in breweries, dairies, soft drinks plants and throughout the processed food industry. Powerful oxidising action also assists stain removal and deodorises. Free-rinsing and non-tainting ensures safe for all food applications.
Low environmental impact, breaks down to materials that are innocuous for waste water treatment.
Caution: Use biocides safely. Always read the label and product information before use.