
5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulphonamide
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:444731-52-6
:
Formula:C₂₁H₂₃N₇O₂S
MDL Number:MFCD11616589
:437.53 g/mol
Melting Pt:285…289 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to light yellow or beige Powder
- Infrared spectrum:Conforms
- Solubility:(0.5 % in DMSO, warmed) Clear solution
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together