L-Biopterin ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22150-76-1
:
Formula:C₉H₁₁N₅O₃
MDL Number:MFCD00036787
:237.2 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together