Order Entry
Poland
ContactUsLinkComponent
Atrazine
Irritant
Health
Environment
Atrazine
Certificates
undefined
Irritant
Health
Environment
Atrazine

Specification Test Results

  • Appearance:
    White to off-white Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    173 to 180 °C
  • Water:
    ≤1 %
  • HPLC:
    ≥95.0 %

Specification Test Results

Frequently Bought Together