Safety ankle boots, lace-up, Compositelite™ Thor, FC11
Supplier: Portwest
Certificates
About this item
Metal-free, anti-static, lightweight boot made from water resistant leather fabric plus a durable, dual-density PU/rubber sole, resistant to oil and heat (300 °C).
- Shock-absorbent heel
- Composite toecap (200 J)
- Puncture resistant composite midsole
Top class design and engineering that offers stylish and durable protection. Non metallic safety components ideal for industrial wear with a strong and lightweight construction.
Colour: Black
Certifications: EN ISO 20345; S3; SRC certified.
Frequently Bought Together