Safety shoes, lace-up, Compositelite™ WORK FC44
: Portwest
Metal-free, anti-static, lightweight safety shoe made from full grain leather.
- Energy absorbing seat region
- Composite toecap (200 J)
- Pierce resistant composite midsole
- Water resistant upper
Hardwearing dual density PU/TPU outsole with superb slip and oil resistance.
: EN ISO 20345:2011, S3, SRC.
Frequently Bought Together