Endurance Hi-Vis earmuffs, PS47
: Portwest
These Easy to clip-on hard hat mounting earmuffs applies the latest standards to deliver the highest level of protection. Comfortable and lightweight, ensure a pleasant wear, even for long periods of use.
- Hi-Visibility colour for visibility in daylight
- Mad of ABS, polystyrene
- Can be adjusted into the standoff position when not in use
- Compatible with PS55, PW98, PS53, PV60, PV64, PV50, PV54, PS50 hard hats
- Excellent attenuation rate
These hearing protection range includes PPE with different level sound attenuation, to be adapted to various working environments giving the correct protection from dangerous noise, without isolating the workers. These updated range of earplugs has best performance level.
: AS/NZS 1270 (SLC(80) 22dB - Class 4); EN 352-3; ANSI S3.19 (NRR 24dB).
: Retail bag which aids presentation for retail sales; Individually packed for vending machines.
Frequently Bought Together