Analytical standard, Robenidine hydrochloride, reference material, Fluka™
Catalog # HONCR1757-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:25875-50-7
- Description:Robenidine hydrochloride
- Description:Robenidine hydrochloride
- Form:Neat