Order Entry
Poland
Orders LinkContactUsLinkComponent
Analytical standard, Fenhexamid, reference material, Fluka™

Analytical standard, Fenhexamid, reference material, Fluka™

Fenhexamid, Reference material, Neat, Glass bottle, 100 mg
Catalog # HONCR1740-100MG
Supplier:   Honeywell Chemicals
Analytical standard, Fenhexamid, reference material, Fluka™
Catalog # HONCR1740-100MG
CAS Number:  
Analytical standard, Fenhexamid, reference material, Fluka™

Fenhexamid, Reference material, Neat, Glass bottle, 100 mg

CAS Number:  
Price
505 zł
Log in for availability
1 * 100 mg
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • Description:
    Fenhexamid
  • Form:
    Neat
  • Cat. No.:
    HONCR1740-100MG
  • Description:
    Fenhexamid
  • CAS No.:
    126833-17-8

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.