Analytical standard, Propargite, reference material, Fluka™
HONCR1712-100MG
:
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:2312-35-8
- Description:Propargite
- Environmentally Preferable:
- Storage temperature:2…8 °C
- Description:Propargite
- Form:Neat