Analytical standard, Abamectin, reference material, Fluka™
Abamectin, Reference material, Neat, Glass bottle, 100 mg
Catalog # HONCR1781-100MG
Supplier: Honeywell Chemicals
Analytical standard, Abamectin, reference material, Fluka™
Catalog # HONCR1781-100MG
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- Description:Abamectin
- Form:Neat
- Cat. No.:HONCR1781-100MG
- Description:Abamectin
- Storage temperature:–20 °C
- CAS No.:71751-41-2



