Analytical standard, Thiabendazol, reference material, Fluka™
Thiabendazol, Reference material, Neat, Glass bottle, 100 mg
Catalog # HONCR1777-100MG
Supplier: Honeywell Chemicals
Analytical standard, Thiabendazol, reference material, Fluka™
Catalog # HONCR1777-100MG
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- Description:Thiabendazol
- Form:Neat
- Cat. No.:HONCR1777-100MG
- Formula:C₁₀H₇N₃S
- Description:Thiabendazol
- CAS No.:148-79-8



