Lithiumbis(trimethylsilyl)amide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4039-32-1
:
Formula:C₆H₁₈LiNSi₂
MDL Number:MFCD00008261
:167.33 g/mol
Boiling Pt:80…84 °C (0.01 mmHg)
Melting Pt:105…107 °C
Density:0.89 g/cm³ (20 °C)
EINECS:223-725-6
ADR:4.3,II
:M-Clarity™ quality level = MQ200
UN:3396
Frequently Bought Together