1,4-Butanesultone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1633-83-6
:
Formula:C₄H₈O₃S
MDL Number:MFCD00006584
:136.17 g/mol
Boiling Pt:153 °C (1013 hPa)
Melting Pt:12.5…15 °C
Density:1.331 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:216-647-9
Frequently Bought Together