Order Entry
Poland
ContactUsLinkComponent
 
 
 
1,3-Propanesultone 97%
 
undefined
 
 
1,3-Propanesultone 97%

 

  • Appearance:
    White to light yellow Low melting solid or liquid
  • Infrared spectrum:
    Conforms
  • Melting point:
    28 to 33 °C
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together