Order Entry
Poland
ContactUsLinkComponent
Cobalt disulphide ≥99.5% (metals basis excluding Ni) (max. 0.2% Ni)
Irritant
Cobalt disulphide ≥99.5% (metals basis excluding Ni) (max. 0.2% Ni)
Catalog # 14005.06
CAS Number:  
Irritant
Cobalt disulphide ≥99.5% (metals basis excluding Ni) (max. 0.2% Ni)
Catalog # 14005.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :123.07 g/mol
Storage Temperature:Ambient
UN:3077
ADR:9,III
 :GHS07
CAS Number:12013-10-4
Formula:CoS₂
MDL Number:MFCD00064735