Titanium(IV) oxide anatase ≥99.7% (metals basis), nanopowder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1317-70-0
:13463-67-7
Formula:TiO₂
MDL Number:MFCD00011269
:79.88 g/mol
Merck Index:14,9472
EINECS:215-280-1
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together