Zinc phosphide, Sigma-Aldrich®
Catalog # SIAL04502-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
About this item
Synonyms: Trizinc diphosphide, Zinc phosphide
:
:258.12 g/mol
EINECS:215-244-5
UN:1714
:GHS02
:GHS06
:GHS09
CAS Number:1314-84-7
Formula:P₂Zn₃
MDL Number:MFCD00049631
Boiling Pt:1100
Melting Pt:420
Density:4.54 g/cm³ (20 °C)
ADR:4.3 I
Merck Index:12,10285
:M-Clarity™ quality level = MQ100