Copper(II) carbonate basic (≥55% Cu)
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 12069-69-1 Molecular Formula: CH2Cu2O5 Formula Weight: 221.11 Physical Form: Powder Density: 4.00 Appearance: Green Melting Point: 200° dec. Merck Reference: 14,2631 MDL No.: MFCD00010976
Synonyms: Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
- CAS Number:12069-69-1
- Formula:CuCO₃·Cu(OH)₂
- MDL Number:MFCD00010976
- Molecular Weight:221.12 g/mol
- Merck Index:14,2631
- Melting Pt:200 °C (dec.)
- Density:4.00
- Storage Temperature:Ambient
- EINECS:235-113-6
- UN:3077
- ADR:9,III
2 Options Available Below
Product Details & Documents
CAS No.: 12069-69-1 Molecular Formula: CH2Cu2O5 Formula Weight: 221.11 Physical Form: Powder Density: 4.00 Appearance: Green Melting Point: 200° dec. Merck Reference: 14,2631 MDL No.: MFCD00010976
Synonyms: Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
- CAS Number:12069-69-1
- Formula:CuCO₃·Cu(OH)₂
- MDL Number:MFCD00010976
- Molecular Weight:221.12 g/mol
- Merck Index:14,2631
- Melting Pt:200 °C (dec.)
- Density:4.00
- Storage Temperature:Ambient
- EINECS:235-113-6
- UN:3077
- ADR:9,III
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



