1,5-Bis(diphenylphosphino)pentane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:27721-02-4
:
Formula:C₂₉H₃₀P₂
MDL Number:MFCD00003052
:440.5 g/mol
Boiling Pt:230 °C (0.1 mmHg)
Melting Pt:43…46 °C
:M-Clarity™ quality level = MQ100
Frequently Bought Together