
Gallium trifluoride ≥99.995% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7783-51-9
:
Formula:F₃Ga
MDL Number:MFCD00011021
:126.72 g/mol
Merck Index:14,4354
Boiling Pt:∼1000 °C
Melting Pt:800 °C (subl.)
Density:4.47
Storage Temperature:Ambient
EINECS:232-004-5
UN:3260
ADR:8,III
:GHS05
Frequently Bought Together