Order Entry
Poland
Orders LinkContactUsLinkComponent
Calcium hypochlorite, tech.

Calcium hypochlorite, tech.

Oxidizing
Corrosive
Irritant
Environment
Calcium hypochlorite, tech.
Certificates
Calcium hypochlorite, tech.
Calcium hypochlorite, tech.

2 Options Available Below

Product Details & Documents

CAS No.: 7778-54-3 Molecular Formula: CaCl2O2 Formula Weight: 142.98 Physical Form: Granular Density: 2.350 Appearance: White to pale brown Odour: Like chlorine pH: 11,5 Melting Point: 100° Merck Reference: 14,1674 UN No.: 1748 MDL No.: MFCD00010900

DeOdourant, oxidizing agent, bleaching agent

  • CAS Number:7778-54-3
  • Formula:CaCl₂O₂
  • MDL Number:MFCD00010900
  • Molecular Weight:142.98 g/mol
  • Merck Index:14,1674
  • Melting Pt:100 °C
  • Density:2.350
  • Storage Temperature:Ambient
  • EINECS:231-908-7
  • UN:1748
  • ADR:5.1,II
Product Family Options

Product Information