Lithium dihydrogen orthophosphate, Sigma-Aldrich®
Catalog # SIAL442682-500G-A
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:103.93 g/mol
EINECS:236-633-6
CAS Number:13453-80-0
Formula:H₂LiO₄P
MDL Number:MFCD00016188