Order Entry
Poland
ContactUsLinkComponent
Isophorone for synthesis, Sigma-Aldrich®
Health
Irritant
Isophorone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Health
Irritant
Isophorone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    14.5 g/l (20 °C)
  • Melting Point:
    -8 °C
  • Molar Mass:
    138.21 g/mol
  • Boiling Point:
    210 - 216 °C (1013 hPa)
  • Vapor Pressure:
    0.33 hPa (20 °C)
  • Flash Point:
    85 °C
  • Refractive Index:
    1.4759 (20 °C, 589 nm)
  • Explosion Limit:
    0.8 - 3.8 %(V)
  • Density:
    0.92 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral,Aqueous solution
  • Ignition Point:
    460 °C
  • Synonyms:
    3,5,5-Trimethyl-2-cyclohexen-1-one
  • Hill Formula:
    C9H14O

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together