


Cyclohexanone oxime 97%
:100-64-1
:
:C₆H₁₁NO
:MFCD00001660
:113.16 g/mol
:206 °C (1013 hPa)
:88…92 °C
:1,13 g/cm³ (20 °C)
:90 °C
:202-874-0
:1325
:GHS02
:GHS07
:GHS08
:Ambient
- Appearance:Off-white to beige to light brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:88 to 92 °C
- Titration with HClO4:≥96.0 %
Frequently Bought Together