Order Entry
Poland
ContactUsLinkComponent
Chloramine-T (sodium salt) trihydrate ≥98% (by iodometry), GPR RECTAPUR®
Irritant
Corrosive
Health
Chloramine-T (sodium salt) trihydrate ≥98% (by iodometry), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Health
Chloramine-T (sodium salt) trihydrate ≥98% (by iodometry), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (iodometric):
    ≥ 98 %
  • Identification (IR):
    Passes test
  • pH (25°C; 5 %):
    8.0 - 11.5
  • Alcohol-insoluble matter:
    ≤ 1.5 %
  • Heavy metals (as Pb):
    ≤ 100 ppm
  • Fe (Iron):
    ≤ 100 ppm
  • Residue insoluble in anhydrous ethanol.:
    Max. 2 %

Specification Test Results

Frequently Bought Together