
Benzenesulphonamide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:98-10-2
:
Formula:C₆H₇NO₂S
MDL Number:MFCD00007930
:157.19 g/mol
Melting Pt:149…152 °C
:M-Clarity™ quality level = MQ200
EINECS:202-637-1
:GHS07
Specification Test Results
- Solubility:4.3 g/l (16 °C)
- Melting Point:149 - 152 °C
- Molar Mass:157.18 g/mol
- Bulk Density:700 kg/m3
- Flash Point:250 °C
Specification Test Results
Related Information
Frequently Bought Together