


4,5-Dichloro-2-nitroaniline ≥98%
Catalog # A17908.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:207.02 g/mol
Melting Pt:180…182 °C
Storage Temperature:Ambient
EINECS:229-657-3
UN:2237
ADR:6.1,III
:GHS06
:GHS07
:GHS09
CAS Number:6641-64-1
Formula:C₆H₄Cl₂N₂O₂
MDL Number:MFCD00007770