Order Entry
Poland
Orders LinkContactUsLinkComponent
MOPS-Na

MOPS-Na

Glass bottle for solids, 100 g
Catalog # MOLE22144595-100G
Marketsource
Supplier:   Molekula
MOPS-Na
Catalog # MOLE22144595-100G
Supplier:  Molekula
Marketsource
CAS Number:  71119-22-7
MOPS-Na

Glass bottle for solids, 100 g

CAS Number:  71119-22-7
Price
645 zł
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt
  • CAS Number:71119-22-7
  • Formula:C₇H₁₄NNaO₄S
  • MDL Number:MFCD00064350
  • Molecular Weight:231.25 g/mol
  • Storage Temperature:Ambient
  • EINECS:428-420-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.