Order Entry
Poland
ContactUsLinkComponent
2'-O-Methyladenosine ≥98.0% (by HPLC, titration analysis)
2'-O-Methyladenosine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAM2291-1G
Supplier:  TCI
CAS Number:  
2'-O-Methyladenosine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAM2291-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    206.0 to 210.0 deg-C
  • Specific rotation [a]20/D:
    -55.0 to -59.0 deg (C=1、H2O)

Specification Test Results

About this item

 :
 :281.27 g/mol
Storage Temperature:Freezer
CAS Number:2140-79-6
Formula:C₁₁H₁₅N₅O₄
MDL Number:MFCD00056002