Order Entry
Poland
ContactUsLinkComponent
Ethyl-1,3-dithiolane-2-carboxylate ≥97.0% (by GC)
Ethyl-1,3-dithiolane-2-carboxylate ≥97.0% (by GC)
Catalog # TCIAD2105-5G
Supplier:  TCI
CAS Number:  
Ethyl-1,3-dithiolane-2-carboxylate ≥97.0% (by GC)
Catalog # TCIAD2105-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :178.26 g/mol
EINECS:243-837-9
CAS Number:20461-99-8
Formula:C₆H₁₀O₂S₂
MDL Number:MFCD00005411