Order Entry
Poland
ContactUsLinkComponent
Isopentyl-4-(dimethylamino)benzoate [contains 2-Methylbutyl 4-(Dimethylamino)benzoate]
Isopentyl-4-(dimethylamino)benzoate [contains 2-Methylbutyl 4-(Dimethylamino)benzoate]
Catalog # TCIAD1760-500G
Supplier:  TCI
CAS Number:  
Isopentyl-4-(dimethylamino)benzoate [contains 2-Methylbutyl 4-(Dimethylamino)benzoate]
Catalog # TCIAD1760-500G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 % (total of isomer)
  • Purity (NMR):
    min. 80 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    33.0 to 38.0 deg-C

Specification Test Results

About this item

 :
 :235.33 g/mol
Merck Index:14,03232
EINECS:244-288-8
CAS Number:21245-01-2
Formula:C₁₄H₂₁NO₂
MDL Number:MFCD00059356