Iron(II) acetate, Sigma-Aldrich®
Catalog # SIAL517933-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:173.94 g/mol
Melting Pt:190 °C
:M-Clarity™ quality level = MQ100
EINECS:221-441-7
CAS Number:3094-87-9
Formula:C₄H₆FeO₄
MDL Number:MFCD00058909