Order Entry
Poland
ContactUsLinkComponent
N-Carbobenzoxy-L-isoleucine ≥98.0% (by HPLC)
N-Carbobenzoxy-L-isoleucine ≥98.0% (by HPLC)
Catalog # TCIAC1354-25G
Supplier:  TCI
CAS Number:  
N-Carbobenzoxy-L-isoleucine ≥98.0% (by HPLC)
Catalog # TCIAC1354-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %
  • Specific rotation [a]20/D:
    +5.0 to +8.0 deg (C=6, EtOH)
  • Solubility in EtOH:
    almost transparency

Specification Test Results

About this item

 :
 :265.31 g/mol
EINECS:221-611-0
CAS Number:3160-59-6
Formula:C₁₄H₁₉NO₄
MDL Number:MFCD00027064